Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMPR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114195 Supplier Invitrogen™ Supplier No. PA5114195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TAMNKHAGGV AEYRASEGKT VEVPYKGDVE NTILDILGGL RSTCTYVGAA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMPR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GMPR
Gene Accession No. P36959
Gene Alias 2310004P21Rik; AV028449; GMP reductase 1; GMPR; GMPR 1; Gmpr1; guanine monophosphate reductase; guanosine 5'-monophosphate oxidoreductase 1; guanosine monophosphate reductase; guanosine monophosphate reductase 1
Gene Symbols GMPR
Host Species Rabbit
Immunogen Immunogen sequence: TAMNKHAGGV AEYRASEGKT VEVPYKGDVE NTILDILGGL RSTCTYVGAA.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2766
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.