Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPR1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982885
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2885 Supplier Bioss Supplier No. BS13462R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.

Specifications

Antigen GMPR1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GMPR
Gene Accession No. P36959
Gene Alias GMP reductase 1; GMPR; GMPR 1; GMPR1; guanine monophosphate reductase; Guanosine 5′-monophosphate oxidoreductase 1; guanosine monophosphate reductase; Guanosine monophosphate reductase 1
Gene Symbols GMPR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GMPR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2766
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.