Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMPR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566960 Supplier Invitrogen™ Supplier No. PA566960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTSCLPALRFI ATPRLSAMPH IDNDVKLDFK D Highest antigen sequence identity to the following orthologs - mouse 44%, rat 44%.

Catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides (PubMed:12009299, PubMed:12669231, PubMed:16359702, PubMed:22037469). Plays a role in modulating cellular differentiation (PubMed:12669231).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMPR2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GMPR2
Gene Accession No. Q9P2T1
Gene Alias 1810008P16Rik; 5730544D12Rik; AA959850; GMP reductase 2; GMPR 2; GMPR2; Guanosine 5'-monophosphate oxidoreductase 2; guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog
Gene Symbols GMPR2
Host Species Rabbit
Immunogen Recombinant Human GMPR2. Recombinant protein control fragment (Product #RP-107604).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51292
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.