Learn More
Invitrogen™ GMPR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551481
Description
Immunogen sequence: YPQLSAVMEC ADAAHGLKGH IISDGGCSCP GDVAKAFGAG ADFVMLGGML AGHSESGGEL IERDGKKYKL FYGMSSEMAM KKYAGGVAEY RASEGKTVEV PFKGDVEHTI RDILGGIRST CTYVGAAKLK ELSRRTTFIR VTQQV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides (PubMed:12009299, PubMed:12669231, PubMed:16359702, PubMed:22037469). Plays a role in modulating cellular differentiation (PubMed:12669231).
Specifications
GMPR2 | |
Polyclonal | |
Unconjugated | |
GMPR2 | |
1810008P16Rik; 5730544D12Rik; AA959850; GMP reductase 2; GMPR 2; GMPR2; Guanosine 5'-monophosphate oxidoreductase 2; guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2T1 | |
GMPR2 | |
Recombinant protein corresponding to Human GMPR2. Recombinant protein control fragment (Product #RP-88934). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.