Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNA12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529928 Supplier Invitrogen™ Supplier No. PA529928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (98%), Rat (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Guanine nucleotide-binding proteins (G proteins) are modulators or transducers in various transmembrane signaling systems. G proteins are composed of 3 subunits; alpha, beta and gamma. The alpha chain contains the guanine nucleotide binding site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNA12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene GNA12
Gene Accession No. Q03113
Gene Alias AI414047; AI504261; G alpha-12; G protein subunit alpha 12; Galpha12; gep; GNA12; Gna-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; guanine nucleotide binding protein, alpha 12; guanine nucleotide-binding protein subunit alpha-12; NNX3; RMP; WUGSC:H_GS165O14.2
Gene Symbols GNA12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human G protein alpha 12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2768
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.