Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNA13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545827
Description
Peptide sequence: SREKTYVKRL VKILLLGAGE SGKSTFLKQM RIIHGQDFDQ RAREEFRPTI.
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems (PubMed:15240885, PubMed:16787920, PubMed:16705036, PubMed:27084452). Activates effector molecule RhoA by binding and activating RhoGEFs (ARHGEF1/p115RhoGEF, ARHGEF11/PDZ-RhoGEF and ARHGEF12/LARG) (PubMed:15240885, PubMed:12515866). GNA13-dependent Rho signaling subsequently regulates transcription factor AP-1 (activating protein-1). Promotes tumor cell invasion and metastasis by activating RhoA/ROCK signaling pathway (PubMed:16787920, PubMed:16705036, PubMed:27084452). Inhibits CDH1-mediated cell adhesion in process independent from Rho activation (PubMed:11976333). [UniProt].
Specifications
GNA13 | |
Polyclonal | |
Unconjugated | |
GNA13 | |
AU024132; AU043124; G alpha 13; G alpha-13; G protein subunit alpha 13; G13; Galpha 13; Galpha13; Gna13; Gna-13; G-protein subunit alpha-13; guanine nucleotide binding protein (G protein), alpha 13; guanine nucleotide binding protein, alpha 13; guanine nucleotide-binding protein subunit alpha-13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10672 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14344 | |
GNA13 | |
Synthetic peptide directed towards the N-terminal of human GNA13 (aa 39-88). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction