Learn More
Invitrogen™ GNA13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117026
Description
Positive Samples: 293T, BT-474, Jurkat, Mouse kidney, Mouse lung, Mouse spleen, Rat kidney Immunogen sequence: MADFLPSRSV LSVCFPGCLL TSGEAEQQRK SKEIDKCLSR EKTYVKRLVK ILLLGAGESG KSTFLKQMRI IHGQDFDQRA REEFRPTIYS NVIKGMRVLV DAREKLHIPW GDNSNQQHGD KMMSFDTRAP MAAQGMVETR VFLQYLPAIR ALWADSGIQN AYDRRREFQL GESVKYFLDN LDKLGEPDYI PSQQDILLAR RPTKGIHEYD.
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems (PubMed:15240885, PubMed:16787920, PubMed:16705036, PubMed:27084452). Activates effector molecule RhoA by binding and activating RhoGEFs (ARHGEF1/p115RhoGEF, ARHGEF11/PDZ-RhoGEF and ARHGEF12/LARG) (PubMed:15240885, PubMed:12515866). GNA13-dependent Rho signaling subsequently regulates transcription factor AP-1 (activating protein-1). Promotes tumor cell invasion and metastasis by activating RhoA/ROCK signaling pathway (PubMed:16787920, PubMed:16705036, PubMed:27084452). Inhibits CDH1-mediated cell adhesion in process independent from Rho activation (PubMed:11976333). [UniProt].
Specifications
| GNA13 | |
| Polyclonal | |
| Unconjugated | |
| GNA13 | |
| AU024132; AU043124; G alpha 13; G alpha-13; G protein subunit alpha 13; G13; Galpha 13; Galpha13; Gna13; Gna-13; G-protein subunit alpha-13; guanine nucleotide binding protein (G protein), alpha 13; guanine nucleotide binding protein, alpha 13; guanine nucleotide-binding protein subunit alpha-13 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10672, 14674, 303634 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.92 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27601, Q14344, Q6Q7Y5 | |
| GNA13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human GNA13 (NP_0065632). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.