Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNA14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587873 Supplier Invitrogen™ Supplier No. PA587873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLGTGESGKS TFIKQMRIIH GSGYSDEDRK GFTKLVYQNI FTAMQAMIRA MDTLRIQYVC EQNKENAQII REVEVDKVSM LSREQVEAIK QLWQDPGIQE CYDRRREYQL SDSAKYYLTD IDRIATPSFV PTQQDVLRVR VPTTGIIEYP FDLENIIFRM VDVGGQRSER RKWIHCFESV TSIIFLVALS EYDQVLAECD NENRMEESKA LFKTIITYPW FLNSSVILFL NKKDLLEEKI MYSHLISYFP EYTGPKQDVR AARDFILKLY QDQNPDKEKV IYSHFTCATD TDNIRFVFAA VKDTILQLNL R; Positive Samples: BxPC-3, 293T, Mouse stomach, Mouse lung, Rat kidney.

This gene encodes a member of the guanine nucleotide-binding, or G protein family. G proteins are heterotrimers consisting of alpha, beta and gamma subunits. The encoded protein is a member of the alpha family of G proteins, more specifically the alpha q subfamily of G proteins. The encoded protein may play a role in pertussis-toxin resistant activation of phospholipase C-beta and its downstream effectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNA14
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gna14
Gene Accession No. O95837, P30677
Gene Alias AU023208; G alpha-14; G protein subunit alpha 14; Gna14; Gna-14; G-protein subunit alpha-14; guanine nucleotide binding protein (G protein), alpha 14; guanine nucleotide binding protein, alpha 14; guanine nucleotide-binding protein 14; guanine nucleotide-binding protein subunit alpha-14; TGFB inducible early growth response 3; Tieg3
Gene Symbols Gna14
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14675, 309242, 9630
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.