Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNA14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597794
Description
GNA14 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the guanine nucleotide-binding, or G protein family. G proteins are heterotrimers consisting of alpha, beta and gamma subunits. The encoded protein is a member of the alpha family of G proteins, more specifically the alpha q subfamily of G proteins. The encoded protein may play a role in pertussis-toxin resistant activation of phospholipase C-beta and its downstream effectors.
Specifications
GNA14 | |
Polyclonal | |
Unconjugated | |
Gna14 | |
AU023208; G alpha-14; G protein subunit alpha 14; Gna14; Gna-14; G-protein subunit alpha-14; guanine nucleotide binding protein (G protein), alpha 14; guanine nucleotide binding protein, alpha 14; guanine nucleotide-binding protein 14; guanine nucleotide-binding protein subunit alpha-14; TGFB inducible early growth response 3; Tieg3 | |
Rabbit | |
Protein G | |
RUO | |
14675, 9630 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O95837, P30677 | |
Gna14 | |
Recombinant Human Guanine nucleotide-binding protein subunit alpha-14 protein (8-303AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction