Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNA15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102572 Supplier Invitrogen™ Supplier No. PA5102572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GNA15.

Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNA15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GNA15
Gene Accession No. O88302, P30678, P30679
Gene Alias Epididymis tissue protein Li 17E; G alpha 15; G alpha 16; g alpha-15; G alpha-16; G protein subunit alpha 15; G[a]15; Galpha15; GNA 15; GNA 16; GNA15; Gna-15; GNA16; G-protein subunit alpha-15; G-protein subunit alpha-16; Gq class; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); guanine nucleotide binding protein, alpha 15; guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16
Gene Symbols GNA15
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNA15(Accession P30679), corresponding to amino acid residues D304-K354.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14676, 2769, 89788
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.