Learn More
Invitrogen™ GNA15 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | GNA15 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GNA15 |
| Gene Accession No. | O88302, P30678, P30679 |
| Gene Alias | Epididymis tissue protein Li 17E; G alpha 15; G alpha 16; g alpha-15; G alpha-16; G protein subunit alpha 15; G[a]15; Galpha15; GNA 15; GNA 16; GNA15; Gna-15; GNA16; G-protein subunit alpha-15; G-protein subunit alpha-16; Gq class; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); guanine nucleotide binding protein, alpha 15; guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16 |
| Gene Symbols | GNA15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.