Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100545 Supplier Invitrogen™ Supplier No. PA5100545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GNAI2.

The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAI2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gnai2
Gene Accession No. P04897, P04899, P08752
Gene Alias adenylate cyclase-inhibiting G alpha protein; BR; C76432; CD49B; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G protein subunit alpha i2; Galphai2; Gia; GIP; GNAI2; Gnai-2; GNAI2B; GPIa; GTP-binding protein (G-alpha-i2); GTP-binding regulatory protein Gi alpha-2 chain; guanidine nucleotide binding protein, (G protein) , alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting activity polypeptide 2; guanine nucleotide-binding protein G(i) subunit alpha-2; guanine nucleotide-binding protein G(i), alpha-2 subunit; H_LUCA15.1; H_LUCA16.1; inhibitory GTP-binding protein subunit alpha 2; VLA-2; VLAA2
Gene Symbols Gnai2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNAI2(Accession P04899), corresponding to amino acid residues N270-I320.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14678, 2771, 81664
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.