Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAI1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA530043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530043 Supplier Thermo Scientific Supplier No. PA530043

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Human (98%), Mouse (98%), Rat (98%), Zebrafish (97%), Xenopus laevis (97%), Pig (98%), Chicken (97%), Bovine (98%), Guinea pig (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GNAI1 Polyclonal antibody specifically detects GNAI1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAI1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene GNAI1
Gene Accession No. P63096
Gene Alias adenylate cyclase-inhibiting G alpha protein; AU046200; BPGTPB; G protein subunit alpha i1; Gi; Gi1 protein alpha subunit; Gi1 protein alpha-subunit; Gialpha1; GNAI1; Gnai-1; GNAI1 protein; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein, alpha inhibiting 1; guanine nucleotide-binding protein G(i) subunit alpha-1; guanine nucleotide-binding protein G(i), alpha-1 subunit; p-alpha-h subunit; unnamed protein product
Gene Symbols GNAI1
Host Species Rabbit
Immunogen Recombinant protein fragment corresponding to a region within amino acids 132 and 318 of Human G protein alpha Inhibitor 1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2770
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.