Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAI3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA527940

Catalog No. PIPA527940


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (88%), Japanese Medaka (91%), Xenopus laevis (93%), Pig (99%), Chicken (97%), Rhesus Monkey (100%), Bovine (98%), Guinea pig (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Heterotrimeric G proteins function to relay information from cell surface receptors to intracellular effectors. Each of a very broad range of receptors specifically detects an extracellular stimulus (a photon, pheromone, odorant, hormone or neurotransmitter) while the effectors (i. e. adenyl cyclase), which act to generate one or more intracellular messengers, are less numerous. In mammals, G protein alpha, beta and gamma polypeptides are encoded by at least 16, 4 and 7 genes, respectively. Most interest in G proteins has been focused on their alpha subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Four distinct classes of Galpha subunits have been identified; these include Gs, Gi, Gq and Galpha 12/13. The Gi class comprises all the known alpha subunits that are susceptible to pertussis toxin modifications, including Galpha i-1, Galpha i-2, Galpha i-3, Galpha o, Galpha t1, Galpha t2, Galpha z and Galpha gust. Of these, the three Galpha i subtypes function to open atrial potassium channels.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GNAI3
Polyclonal
Unconjugated
GNAI3
87U6; AI158965; ARCND1; AW537698; G protein subunit alpha i3; g(i) alpha-3; Galphai3; GIP3A; GNAI3; Gnai-3; guanine nucleotide binding protein (G protein), alpha inhibiting 3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; guanine nucleotide binding protein, alpha inhibiting 3; Guanine nucleotide binding, protein, alpha inhibiting polypeptide 3; Guanine nucleotide-binding protein G(i) subunit alpha; guanine nucleotide-binding protein G(k) subunit alpha
Rabbit
Antigen affinity chromatography
RUO
14679, 2773
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.94 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P08754, Q9DC51
GNAI3
Recombinant fragment corresponding to a region within amino acids 68 and 354 of Human G protein alpha Inhibitor 3
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.