Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNAI3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30390020UL
Description
GNAI3 Polyclonal specifically detects GNAI3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GNAI3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
FLJ26559, G(i) alpha-3, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 3,87U6, guanine nucleotide-binding protein G(k) subunit alpha | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GNAI3 (NP_006487.1). | |
20 μL | |
Signal Transduction | |
2773 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction