Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527964 Supplier Invitrogen™ Supplier No. PA527964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (89%), Xenopus laevis (92%), Chicken (95%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems. G(olf) alpha mediates signal transduction within the olfactory neuroepithelium and the basal ganglia. May be involved in some aspect of visual transduction, and in mediating the effect of one or more hormones/neurotransmitters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Gnal
Gene Accession No. P38405, Q8CGK7
Gene Alias 2610011C15Rik; 9630020G10Rik; adenylate cyclase-stimulating G alpha protein, olfactory type; DYT25; G alpha 10; G protein subunit alpha L; Galphaolf; Gna10; Gnal; Golf; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide, olfactory type; guanine nucleotide binding protein, alpha 10; guanine nucleotide binding protein, alpha stimulating, olfactory type; guanine nucleotide-binding protein G(olf) subunit alpha; guanine nucleotide-binding protein G(olf), alpha subunit; Olf; olfa; Olf-alpha protein (olfactory neuron-specific G protein); RGD1305940
Gene Symbols Gnal
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GNAL. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14680, 2774
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.