Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597795
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597795 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597795 Supplier Invitrogen™ Supplier No. PA597795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNAL Polyclonal Antibody for Western Blot, IHC (P), ELISA

Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems. G(olf) alpha mediates signal transduction within the olfactory neuroepithelium and the basal ganglia. May be involved in some aspect of visual transduction, and in mediating the effect of one or more hormones/neurotransmitters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.725 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Gnal
Gene Accession No. P38405
Gene Alias 2610011C15Rik; 9630020G10Rik; adenylate cyclase-stimulating G alpha protein, olfactory type; DYT25; G alpha 10; G protein subunit alpha L; Galphaolf; Gna10; Gnal; Golf; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide, olfactory type; guanine nucleotide binding protein, alpha 10; guanine nucleotide binding protein, alpha stimulating, olfactory type; guanine nucleotide-binding protein G(olf) subunit alpha; guanine nucleotide-binding protein G(olf), alpha subunit; Olf; olfa; Olf-alpha protein (olfactory neuron-specific G protein); RGD1305940
Gene Symbols Gnal
Host Species Rabbit
Immunogen Recombinant Human Guanine nucleotide-binding protein G(olf) subunit alpha protein (4-116AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2774
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.