Learn More
Invitrogen™ GNAO1 Polyclonal Antibody

Description
This antibody is predicted to react with bovine based on sequence homology.
Specifications
Specifications
| Antigen | GNAO1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GNAO1 |
| Gene Accession No. | P09471, P18872, P59215 |
| Gene Alias | alphaO; AW050213; DKFZp686O0962; EIEE17; G alpha o; G protein subunit alpha o1; Galphao; G-ALPHA-o; Gna0; Gnao; GNAO 1; Gnao1; Go alpha; GO2-q chimeric G-protein; GTP-binding protein alpha o; GTP-binding protein alpha-o subunit; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide O; guanine nucleotide binding protein, alpha O; guanine nucleotide binding protein, alpha O polypeptide 1; guanine nucleotide-binding protein G(o) subunit alpha; guanine nucleotide-binding regulatory protein 2; HLA-DQB1; RATBPGTPC |
| Gene Symbols | GNAO1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.