Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530044 Supplier Invitrogen™ Supplier No. PA530044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, mouse lung, rat brain, rat lung. Predicted reactivity: Mouse (97%), Rat (98%), Xenopus laevis (98%), Chicken (98%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Heterotrimeric G proteins are membrane bound GTPases that are linked to 7-TM receptors. Each G protein contains an alpha-, beta- and gamma-subunit and is bound to GDP in the 'off' state. Ligand-receptor binding results in detachment of the G protein, switching it to an 'on' state and permitting Galpha activation of second messenger signalling cascades. There are several types of Galpha proteins; Galphas and Galphai bind directly to adenylyl cyclase and stimulate or inhibit its activity, leading to an increase or decrease in cAMP levels respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAO1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene GNAO1
Gene Accession No. P09471, P18872, P59215
Gene Alias alphaO; AW050213; DKFZp686O0962; EIEE17; G alpha o; G protein subunit alpha o1; Galphao; G-ALPHA-o; Gna0; Gnao; GNAO 1; Gnao1; Go alpha; GO2-q chimeric G-protein; GTP-binding protein alpha o; GTP-binding protein alpha-o subunit; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide O; guanine nucleotide binding protein, alpha O; guanine nucleotide binding protein, alpha O polypeptide 1; guanine nucleotide-binding protein G(o) subunit alpha; guanine nucleotide-binding regulatory protein 2; HLA-DQB1; RATBPGTPC
Gene Symbols GNAO1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GNAO1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14681, 2775, 50664
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.