Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNAQ Antibody (GNAQ/2434), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308585PE
Description
GNAQ Monoclonal antibody specifically detects GNAQ in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
GNAQ | |
Monoclonal | |
PE | |
GAQG-ALPHA-q, guanine nucleotide binding protein (G protein), q polypeptide, Guanine nucleotide-binding protein alpha-q, guanine nucleotide-binding protein G(q) subunit alpha | |
Recombinant full-length human GNAQ protein (Uniprot: P50148) | |
100 μg | |
Cardiovascular Biology | |
2776 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry, ELISA, Immunofluorescence | |
GNAQ/2434 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction