Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNAQ Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131985
Description
GNAQ Polyclonal specifically detects GNAQ in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GNAQ | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
GNAQ | |
Recombinant protein encompassing a sequence within the center region of human GNAQ. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
GAQG-ALPHA-q, guanine nucleotide binding protein (G protein), q polypeptide, Guanine nucleotide-binding protein alpha-q, guanine nucleotide-binding protein G(q) subunit alpha | |
Rabbit | |
42 kDa | |
0.1 mL | |
Cardiovascular Biology | |
2776 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction