Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GNAQ, Mouse, Unlabeled, Clone: 8A11.1,

Catalog No. MABS1135MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1135MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1135MI Supplier MilliporeSigma™ Supplier No. MABS1135
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Guanine nucleotide-binding protein G(q) subunit alpha (UniProt P50148; also known as Guanine nucleotide-binding protein alpha-q) is encoded by the GNAQ (also known as GAQ, SWS) gene (Gene ID 2776) in human. Heterotrimeric G proteins are activated by G protein-coupled receptors (GPCRs) and are composed of alpha (α), beta (β), and gamma (γ) subunits. There exists also monomeric Ras superfamily of small GTPases that are commonly referred to as small G-proteins. G-proteins mediate signal transduction initiated by GPCRs. Ligand binding activates GPCRs by inducing a conformational change. The activated GPCRs catalyzes the GDP-to-GTP exchange of the Gα subunit, causing a dissociation of the Gβγ dimer from the now GTP-bound Gα subunit. The Gα subunit gradually hydrolyzes the bound GTP to GDP and eventually re-associates with Gβγ and re-starts the cycle. There exist at least 20 different Gα subunits, which are separated into four main groups, Gi, Gs, Gq, and G12/13. Four different genes code for Gq alpha subunits, GNAQ, GNA11, GNA14, and GNA15. Gq proteins mediate a variety of cellular signaling events by activating phospholipase C (PLC). Gq-coupled GPCRs include α1-adrenoreceptor, angiotensin II receptor type 1, calcitonin receptor, muscarinic receptors (M1, M3, and M5), histamine H1 receptor, Serotonin 5-HT2 receptors, vasopressin type 1 receptors 1A and 1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAQ
Applications Western Blot
Classification Monoclonal
Clone 8A11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene GNAQ, GAQ, SWS
Gene Accession No. P50148
Host Species Mouse
Immunogen GST-tagged recombinant internal fragment of human GNAQ.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002063
Test Specificity Clone 8A11.1 was raised against a human Gqα (GNAQ) recombinant fragment. Clone 8A11.1 reactivity toward Gq/11α (GNA11), Gq/14α (GNA14), and Gq/15α (GNA15) has not been tested.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.