Learn More
Invitrogen™ GNAS Polyclonal Antibody

Description
Recommended positive controls: HepG2, Raji, mouse brain, NIH-3T3, JC, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (97%), Xenopus Tropicalis (97%), Dog (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | GNAS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | GNAS |
| Gene Accession No. | O95467, P63092, P63094, P84996, Q5JWF2, Q63803, Q6R0H6, Q6R0H7, Q9Z0F1 |
| Gene Alias | 5530400H20Rik; A930027G11Rik; Adenylate cyclase-stimulating G alpha protein; AHO; ALEX; GNAS; NESP55; alpha-stimulatory subunit of GTP-binding protein; alternative gene product encoded by XL-exon; C130027O20Rik; C20orf45; dJ309F20.1.1; dJ806M20.3.3; Extra large alphas protein; G protein alpha subunit; G protein subunit alpha S; GAIPIRRH peptide; G-alpha-8; Galphas; Gnas; GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus; gnas {ECO:0000312; GNAS complex locus; GNAS complex locus; SCG6 (secretogranin VI); GNAS1; Gnasxl; Gnpas; GPIPIRRH peptide; GPSA; Gs alpha subunit; GSA; G-SALPHA60A; GSP; GTP-binding regulatory protein (G-s-alpha).; GTP-binding regulatory protein alpha subunit exon 1; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide binding protein alpha s; guanine nucleotide binding protein alpha subunit; guanine nucleotide binding protein, alpha stimulating activity polypeptide 1; guanine nucleotide regulatory protein; guanine nucleotide-binding protein G(s) subunit alpha; guanine nucleotide-binding protein G(s) subunit alpha isoforms; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; guanine nucleotide-binding protein G(s) subunit alpha; guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; guanine nucleotide-binding protein G(s) subunit alpha; SCG6 (secretogranin VI); guanine nucleotide-binding protein G(s), alpha subunit; guanine nucleotide-binding protein Gs alpha1; guanine nucleotide-binding protein G-s, alpha subunit; guanine nucleotide-binding protein, alpha-stimulating activity polypeptide 1; LHAL tetrapeptide; LSAL tetrapeptide; MGC33735; MNCb-5546; Nesp; NESP55; Nespl; neuroendocrine secretory protein; neuroendocrine secretory protein 55; Oedsml; Oed-Sml; OTTHUMP00000031742; OTTHUMP00000196026; OTTHUMP00000196030; P1; P2; P3; PHP1A; PHP1B; PHP1C; POH; PORGSA1; Protein ALEX; protein ALEX; protein GNAS; protein NESP55; protein ALEX; protein GNAS; protein SCG6 (secretogranin VI); RGD:2716}; RP4-543J19.4; SCG6; secretogranin VI; SgVI; stimulatory G-protein alpha subunit; stimulatory GTP binding protein; unnamed protein product; XLalphas; XLas |
| Gene Symbols | GNAS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.