Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GNAT1, Mouse, Unlabeled, Clone: 11A8.1,

Catalog No. MABN1578MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1578MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1578MI Supplier MilliporeSigma™ Supplier No. MABN1578
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Guanine nucleotide-binding protein G(t) subunit alpha-1 (UniProt P11488; also known as Transducin alpha-1 chain) is encoded by the GNAT1 (also known as GNATR, CSNB1G, CSNBAD3) gene (Gene ID 2779) in human. Heterotrimeric G proteins are activated by G protein-coupled receptors (GPCRs) and are composed of alpha (α), beta (β), and gamma (γ) subunits. There exists also monomeric Ras superfamily of small GTPases that are commonly referred to as small G-proteins. G-proteins mediate signal transduction initiated by GPCRs. Ligand binding activates GPCRs by inducing a conformational change. The activated GPCRs catalyze the GDP-to-GTP exchange of the Gα subunit, causing a dissociation of the Gβγ dimer from the now GTP-bound Gα subunit. The Gα subunit gradually hydrolyzes the bound GTP to GDP and eventually re-associates with Gβγ and re-starts the cycle. There exist at least 20 different Gα subunits, which are separated into four main groups, Gi, Gs, Gq, and G12/13. Eight different genes code for Gi alpha subunits, including GNAT1 and GNAT2 that code for Gt in the rod (rTα by GNAT1) and cone (cTα by GNAT2) cells of the retina, where they function as the alpha subunit of the trimeric G protein transducin. The α-subunit binds GTP and activates cyclic GMP phosphodiesterase (PDE), whereas the β and γ subunits mediate interaction with rhodopsin. Mutations in RHO, PDE6B, and GNAT1 genes are associated with autosomal dominant congenital stationary night blindness (CSNB), and patients with mutations in these genes exhibit the Riggs- or Schubert-Bornschein-type electronegative electroretinogram (ERG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAT1
Applications Western Blot
Classification Monoclonal
Clone 11A8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene GNAT1, GNATR, CSNB1G, CSNBAD3
Gene Accession No. P11488
Host Species Mouse
Immunogen GST-tagged recombinant human GNAT1 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000163
Test Specificity Clone 11A8.1 was raised against human Gt alpha-1 subunit (GNAT1) recombinant fragment. Cross-reactivity toward human Gt alpha-2 subunit (GNAT2) has not been determined.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.