Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1724

Catalog No. PA1724


Only null left
Edge
Add to Cart

Description

Description

PA1-724 detects rod transducin alpha (Tr alpha) from bovine outer rod segments and sheep retinal extract. PA1-724 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~38 kDa protein representing Tr alpha from sheep retinal extract. The PA1-724 immunizing peptide corresponds to amino acid residues 18-31 from human Tr alpha. This peptide (Cat. # PEP-153) is available for use in neutralization and control experiments.

Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phosphodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in rods. This gene is also expressed in other cells, and has been implicated in bitter taste transduction in rat taste cells. Mutations in this gene result in autosomal dominant congenital stationary night blindness. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GNAT1
Polyclonal
Unconjugated
GNAT1
CSNB1G; CSNBAD3; G protein subunit alpha transducin 1; GBT1; Gnat1; Gnat-1; GNATR; guanine nucleotide binding protein (G protein), alpha transducing 1; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1; guanine nucleotide binding protein, alpha transducing 1; guanine nucleotide-binding protein G(t) subunit alpha-1; guanine nucleotide-binding protein G(T), alpha-1 subunit; Ird1; Ird2; irdc; irdr; rod-type transducin alpha subunit; Tr alpha; Tralpha; transducin; transducin alpha subunit; transducin alpha-1 chain; transducin alpha-subunit; transducin, rod-specific; transducin-alpha
Rabbit
Antigen Affinity Chromatography
RUO
101121976, 2779, 281794
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P04695, P11488
GNAT1
Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C.
100 μg
Primary
Human, Bovine, Sheep
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.