Learn More
Invitrogen™ GNAT1 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse and Xenopus based on sequence homology.
Specifications
Specifications
| Antigen | GNAT1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GNAT1 |
| Gene Accession No. | P11488, P20612 |
| Gene Alias | CSNB1G; CSNBAD3; G protein subunit alpha transducin 1; GBT1; Gnat1; Gnat-1; GNATR; guanine nucleotide binding protein (G protein), alpha transducing 1; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1; guanine nucleotide binding protein, alpha transducing 1; guanine nucleotide-binding protein G(t) subunit alpha-1; guanine nucleotide-binding protein G(T), alpha-1 subunit; Ird1; Ird2; irdc; irdr; rod-type transducin alpha subunit; Tr alpha; Tralpha; transducin; transducin alpha subunit; transducin alpha-1 chain; transducin alpha-subunit; transducin, rod-specific; transducin-alpha |
| Gene Symbols | GNAT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.