Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587834 Supplier Invitrogen™ Supplier No. PA587834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGSGASAEDK ELAKRSKELE KKLQEDADKE AKTVKLLLLG AGESGKSTIV KQMKIIHQDG YSPEECLEFK AIIYGNVLQS ILAIIRAMTT LGIDYAEPSC ADDGRQLNNL ADSIEEGTMP PELVEVIRRL WKDGGVQACF ERAAEYQLND SASYYLNQLE RITDPEYLPS EQDVLRSRVK TTGIIETKFS VKDLNFRMFD VGGQRSERKK WIHCFEGVTC; Positive Samples: HepG2.

Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gnat2
Gene Accession No. P19087
Gene Alias ACHM4; AW490837; cone-type transducin alpha subunit; G protein subunit alpha transducin 2; Gnat2; Gnat-2; GNATC; Gt-2; guanine nucleotide binding protein (G protein), alpha transducing 2; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; guanine nucleotide binding protein, alpha transducing 2; guanine nucleotide-binding protein G(t) subunit alpha-2; RP5-1160K1.4; Tcalpha; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide
Gene Symbols Gnat2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human GNAT2 (NP_0052631).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2780
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.