Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119558
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones.
Specifications
| GNAT2 | |
| Polyclonal | |
| Unconjugated | |
| Gnat2 | |
| ACHM4; AW490837; cone-type transducin alpha subunit; G protein subunit alpha transducin 2; Gnat2; Gnat-2; GNATC; Gt-2; guanine nucleotide binding protein (G protein), alpha transducing 2; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; guanine nucleotide binding protein, alpha transducing 2; guanine nucleotide-binding protein G(t) subunit alpha-2; RP5-1160K1.4; Tcalpha; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14686, 2780, 365901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.92 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P19087, P50149 | |
| Gnat2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human GNAT2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction