Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNAT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GNAT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GNAT2 Polyclonal specifically detects GNAT2 in Human samples. It is validated for Western Blot.Specifications
GNAT2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2780 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
ACHM4guanine nucleotide-binding protein G(t) subunit alpha-2, cone-type transducin alpha subunit, GNATC, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 2, guanine nucleotide-binding protein G(t), alpha-2 subunit, Transducin alpha-2 chain, transducin, cone-specific, alpha polypeptide | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human GNAT2 (NP_005263.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title