Learn More
Invitrogen™ GNAT3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total GNAT3 protein.
Specifications
Specifications
| Antigen | GNAT3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | GNAT3 |
| Gene Accession No. | A8MTJ3, P29348, Q3V3I2 |
| Gene Alias | alpha-gustducin; G protein subunit alpha transducin 3; GDCA; Ggust; Gnat3; gnat-3; Gtn; guanine nucleotide binding protein, alpha transducing 3; guanine nucleotide-binding protein G(t) subunit alpha-3; gustatory G protein; gustducin alpha-3 chain; gustducin, alpha polypeptide |
| Gene Symbols | GNAT3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.