Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1203433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203433 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203433 Supplier Novus Biologicals Supplier No. NB1203433

Rabbit Polyclonal Antibody

GNB1 Polyclonal specifically detects GNB1 in Human, Mouse, Bovine, Drosophila, Sheep samples. It is validated for Western Blot.

Specifications

Antigen GNB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias beta subunit, signal-transducing proteins GS/GI, G protein, beta-1 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1, Transducin beta chain 1
Gene Symbols GNB1
Host Species Rabbit
Immunogen Synthetic Peptide: R(8) Q E A E Q L K N Q I R D A R K A C(25)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2782
Test Specificity The NB120-3433 immunizing peptide corresponds to amino acid residues 8-25 from human Tr beta.
Target Species Human, Mouse, Bovine, Drosophila, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.