Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216699
Description
GNB1 Polyclonal specifically detects GNB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
GNB1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
GNB1 | |
Recombinant protein encompassing a sequence within the center region of human GNB1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
2782 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Proximity Ligation Assay | |
beta subunit, signal-transducing proteins GS/GI, G protein, beta-1 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1, Transducin beta chain 1 | |
Rabbit | |
37 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Expected to cross react based on sequence identity: Chicken (96%), Dog (100%), Rat (100%), Xenopus laevis (96%), Bovine (100%), Xenopus tropicalis (96%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction