Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gnb1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1725
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1725 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1725 Supplier Invitrogen™ Supplier No. PA1725
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-725 detects transducin beta (T beta) from sheep, bovine, mouse and drosophila. PA1-725 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~37 kDa protein representing Tr beta from sheep and bovine retinal and optic nerve extracts as well as mouse eye samples and drosophila. The PA1-725 immunizing peptide corresponds to amino acid residues 8-25 from human Tr beta. This sequence is completely conserved between human, amphibian, bovine and rat. PA1-725 immunizing peptide (Cat. # PEP-154) is available for use in neutralization and control experiments.

Vision involves the conversion of light into electrochemical signals that are processed by the retina and subsequently sent to and interpreted by the brain. The process of converting light into an electrochemical signal begins when the membrane-bound protein, rhodopsin, absorbs light within the retina. Photoexcitation of rhodopsin causes the cytoplasmic surface of the protein to become catalytically active. In the active state, rhodopsin activates transducin, a GTP binding protein. Once activated, transducin promotes the hydrolysis of cGMP by phosphodiesterase (PDE). The decrease of intracellular cGMP concentration causes the ion channels within the outer segment of the rod or cone to close, thus causing membrane hyperpolarization and, eventually, signal transmission. Rhodopsin activity is believed to be shut off by phosphorylation followed by binding of the soluble protein, arrestin. Transducin, once activated by rhodopsin, promotes the hydrolysis of cGMP by PDE. The subunit composition of transducin differs between different photoreceptor cells. Rod transducin consists of rod transducin alpha (Tr alpha), T beta, and T gamma. Cone transducin is composed of cone transducin alpha (Tc alpha), T beta and T gamma. Differential transducin subunit composition of transducin is believed to be responsible for the different light sensitivities between photoreceptive cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gnb1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene GNB1
Gene Accession No. P54311, P62871, P62873, P62874
Gene Alias AA409223; beta subunit, signal-transducing proteins GS/GI; C77571; G protein subunit beta 1; gbb1; GNB1; Gnb-1; gnb1 protein; gnb1a; guanine nucleotide binding protein (G protein), beta 1; guanine nucleotide binding protein (G protein), beta polypeptide 1; guanine nucleotide binding protein (G protein), beta polypeptide 1a; guanine nucleotide binding protein, beta 1; Guanine nucleotide-binding protein beta 1; guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1; guanine nucleotide-binding protein, beta-1 subunit; MRD42; T beta; testicular tissue protein Li 72; transducin beta chain 1; transducin beta subunit; unnamed protein product; wu:fb50b09; wu:fj94h04
Gene Symbols GNB1
Host Species Rabbit
Immunogen Synthetic Peptide: R(8) Q E A E Q L K N Q I R D A R K A C(25).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101104166, 14688, 24400, 2782, 281201
Target Species Human, Mouse, Rat, Bovine, Sheep, Drosophila
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.