Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNB1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598869
Description
GNB1L Polyclonal Antibody for Western Blot, ELISA
GNB1L is a G-protein beta-subunit-like polypeptide which is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 6 WD repeats and is highly expressed in the heart. Therefore, this gene may contribute to the etiology of those disorders.This gene encodes a G-protein beta-subunit-like polypeptide which is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 6 WD repeats and is highly expressed in the heart. The gene maps to the region on chromosome 22q11, which is deleted in DiGeorge syndrome, trisomic in derivative 22 syndrome and tetrasomic in cat-eye syndrome. Therefore, this gene may contribute to the etiology of those disorders.
Specifications
GNB1L | |
Polyclonal | |
Unconjugated | |
GNB1L | |
DGCRK3; Estm55; FKSG1; G protein subunit beta 1 like; g protein subunit beta-like protein 1; Gm16314; Gnb1l; G-protein beta subunit-like protein; guanine nucleotide binding protein (G protein), beta polypeptide 1-like; guanine nucleotide binding protein beta-subunit-like polypeptide; guanine nucleotide-binding protein subunit beta-like protein 1; GY2; KIAA1645; Me49f07; OTTMUSG00000033458; WD repeat-containing protein 14; WD40 repeat-containing protein deleted in VCFS; Wdr14; WDVCF; zgc:110763 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54584 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BYB4 | |
GNB1L | |
Recombinant Human Guanine nucleotide-binding protein subunit beta-like protein 1 protein (1-327AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction