Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216700
Description
GNB2 Polyclonal specifically detects GNB2 in Human, Mouse samples. It is validated for Western Blot.Specifications
GNB2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
beta-2 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 2, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 2, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2, signal-transducing guanine nucleotide-binding regulatory protein beta subunit | |
Rabbit | |
37 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Expected to cross react based on sequence identity: Rat (100%), Zebrafish (91%), Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.5mg/mL | |
Western Blot 1:1000-1:10000 | |
P62879 | |
GNB2 | |
Recombinant protein encompassing a sequence within the center region of human GNB2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
2783 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction