Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNB2 Recombinant Rabbit Monoclonal Antibody (6U9B3)
GREENER_CHOICE

Catalog No. PIMA535827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535827 Supplier Invitrogen™ Supplier No. MA535827
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MSELEQLRQE AEQLRNQIRD ARKACGDSTL TQITAGLDPV GRIQMRTRRT LRGHLAKIYA MHWGTDSRLL VSASQDGKLI IWDSYTTNKV.

Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. This gene contains a trinucleotide (CCG) repeat length polymorphism in its 5' UTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNB2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 6U9B3
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gnb2
Gene Accession No. P54313, P62879, P62880
Gene Alias G protein subunit beta 2; g protein subunit beta-2; G protein, beta-2 subunit; Gnb2; Gnb-2; guanine nucleotide binding protein (G protein), beta 2; guanine nucleotide binding protein (G protein), beta polypeptide 2; guanine nucleotide binding protein beta 2 subunit; guanine nucleotide binding protein, beta 2; guanine nucleotide binding protein, beta polypeptide 2; guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 2; guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2; guanine nucleotide-binding protein, beta 2; guanine nucleotide-binding protein, beta-2 subunit; signal-transducing guanine nucleotide-binding regulatory protein beta subunit; transducin beta chain 2
Gene Symbols Gnb2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human GNB2 (P62879).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14693, 2783, 81667
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.