Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB3 Antibody, Novus Biologicals™
SDP

Catalog No. p-7199998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017941 0.025 mL
NB10098705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017941 Supplier Novus Biologicals Supplier No. NB100987050.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNB3 Polyclonal specifically detects GNB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen GNB3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1: 300 - 1: 2000, Immunohistochemistry 1: 300 - 1: 2000, Immunohistochemistry-Paraffin 1: 300- 1: 2000, Immunohistochemistry-Frozen 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Accession No. P16520
Gene Alias G protein, beta-3 subunit, GTP-binding regulatory protein beta-3 chain, guanine nucleotide binding protein (G protein), beta polypeptide 3, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 3, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-3, hypertension associated protein, Transducin beta chain 3
Gene Symbols GNB3
Host Species Rabbit
Immunogen A synthetic peptide from the n-terminal of human GBB3 conjugated to an immunogenic carrier protein was used as the immunogen. The peptide is homologous in rat and mouse.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2784
Test Specificity GBB3.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.