Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNB3/GNB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519401
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519401 Supplier Invitrogen™ Supplier No. PA519401
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

Heterotrimeric guanine nucleotide-binding proteins in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNB3/GNB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GNB3
Gene Accession No. P16520, Q9HAV0
Gene Alias 6720453A21Rik; CMTD1F; CSNB1H; G protein beta-4 subunit; G protein beta-subunit; G protein subunit beta 3; G protein subunit beta 4; G protein, beta-3 subunit; G(beta)4; gbb3; gbb4; Gnb3; Gnb-3; GNB4; G-protein beta 4 subunit; GTP-binding regulatory protein beta-3 chain; guanine nucleotide binding protein (G protein), beta 3; guanine nucleotide binding protein (G protein), beta 4; guanine nucleotide binding protein (G protein), beta polypeptide 3; guanine nucleotide binding protein (G protein), beta polypeptide 4; guanine nucleotide binding protein beta 4 subunit; guanine nucleotide binding protein, beta 3; guanine nucleotide binding protein, beta 4; guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 3; guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-3; Guanine nucleotide-binding protein subunit beta-4; guanine nucleotide-binding protein, beta-3 subunit; hypertension associated protein; transducin beta chain 3; Transducin beta chain 4
Gene Symbols GNB3, GNB4
Host Species Goat
Immunogen Synthetic peptide sequence (SGHDNRVSCLGVT) corresponding to the internal amino acids of GNB3 (aa 308-321).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2784, 59345
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.