Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142216
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142216 Supplier Invitrogen™ Supplier No. PA5142216
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: No cross-reactivity expected with GNB1, 2 and 4. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.

Heterotrimeric guanine nucleotide-binding proteins in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNB3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GNB3
Gene Accession No. P16520
Gene Alias CSNB1H; G protein beta-subunit; G protein subunit beta 3; G protein, beta-3 subunit; gbb3; GNB3; Gnb-3; GTP-binding regulatory protein beta-3 chain; guanine nucleotide binding protein (G protein), beta 3; guanine nucleotide binding protein (G protein), beta polypeptide 3; guanine nucleotide binding protein, beta 3; guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 3; guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-3; guanine nucleotide-binding protein, beta-3 subunit; hypertension associated protein; transducin beta chain 3
Gene Symbols GNB3
Host Species Goat
Immunogen Peptide with sequence DCMSLAVSPDFN (aa 186-197).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2784
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.