Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113327
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113327 Supplier Invitrogen™ Supplier No. PA5113327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNB4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNB4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Gnb4
Gene Accession No. O35353, P29387, Q9HAV0
Gene Alias 6720453A21Rik; CMTD1F; G protein beta-4 subunit; G protein subunit beta 4; G(beta)4; gbb4; Gnb4; G-protein beta 4 subunit; guanine nucleotide binding protein (G protein), beta 4; guanine nucleotide binding protein (G protein), beta polypeptide 4; guanine nucleotide binding protein beta 4 subunit; guanine nucleotide binding protein, beta 4; guanine nucleotide-binding protein subunit beta-4; Transducin beta chain 4
Gene Symbols Gnb4
Host Species Rabbit
Immunogen Recombinant Human Guanine nucleotide-binding protein subunit beta-4 protein (105-185AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14696, 294962, 59345
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.