Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119972
This item is not returnable.
View return policy
Description
Positive Control: Human placenta tissue lysate, 293 cell lysate, A549 cell lysate, mouse brain lysates, rat brain lysates. Subcellular Location: Cytosol, extracellular exosome, lysosomal membrane, heterotrimeric G-protein complex, cytoplasm.
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors.
Specifications
| GNB4 | |
| Polyclonal | |
| Unconjugated | |
| Gnb4 | |
| 6720453A21Rik; CMTD1F; G protein beta-4 subunit; G protein subunit beta 4; G(beta)4; gbb4; Gnb4; G-protein beta 4 subunit; guanine nucleotide binding protein (G protein), beta 4; guanine nucleotide binding protein (G protein), beta polypeptide 4; guanine nucleotide binding protein beta 4 subunit; guanine nucleotide binding protein, beta 4; guanine nucleotide-binding protein subunit beta-4; Transducin beta chain 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14696, 294962, 59345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O35353, P29387, Q9HAV0 | |
| Gnb4 | |
| Recombinant protein within human GNB4 aa 1-150. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction