Learn More
Invitrogen™ GNB5 Polyclonal Antibody

Description
Antibody detects endogenous levels of total GNB5.
Specifications
Specifications
| Antigen | GNB5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Gnb5 |
| Gene Accession No. | O14775, P62881, P62882 |
| Gene Alias | flail; flailer; flr; G protein subunit beta 5; G protein, beta subunit 5L; G protein, beta-5 subunit; GB5; GBB5; Gbeta5; GBS; Gnb5; guanine nucleotide binding protein (G protein), beta 5; guanine nucleotide binding protein (G protein), beta polypeptide 5; guanine nucleotide binding protein, beta 5; guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; guanine nucleotide-binding protein, beta-5 subunit; hug; transducin beta chain 5 |
| Gene Symbols | Gnb5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.