Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GNE 9278
GREENER_CHOICESDP

Catalog No. 636950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-695-0 50 mg
63-691-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-695-0 Supplier Tocris Bioscience™ Supplier No. 6369/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of NMDA receptors; acts in transmembrane domain

GNE 9278 is a positive allosteric modulator of NMDA receptors. Potentiates NMDA receptors containing GluN2A, 2B, 2C and 2D subunits (EC50 values are 0.74, 3.07, 0.47 and 0.32 μM in a Ca2+ influx assay, respectively). Binds within the transmembrane domain and displays preference for agonist-bound NMDA receptors. Slows receptor deactivation and enhances the potency of glutamate and glycine .

Specifications

Quantity 50 mg
Molecular Formula C21H27N5O3S
Formula Weight 429.54
Product Type Positive Allosteric Modulator
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.