Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552380 Supplier Invitrogen™ Supplier No. PA552380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDFDINTRLH TIVRGEDEAA MVESVGLALV KLPDVLNRLK PDIMIVHGDR FDALALATSA ALMNIRILHI EGGEVSGTID DSIRHAITKL AHYHVCCTRS AEQHLISMCE DHDRILLAGC PSYDKLLSAK NKD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

GNE is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of malignant cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNE
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GNE
Gene Accession No. Q9Y223
Gene Alias 2310066H07Rik; bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; DMRV; GLCNE; glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase; GNE; IBM2; ManAc kinase; N-acetylmannosamine kinase; N-acylmannosamine kinase; NM; Uae1; UDP-GlcNAc-2-epimerase; UDP-GlcNAc-2-epimerase/ManAc kinase; UDP-N-acetylglucosamine 2-epimerase (hydrolyzing); UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase; Uridine diphosphate-N-acetylglucosamine-2-epimerase
Gene Symbols GNE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GNE. Recombinant protein control fragment (Product #RP-89764).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.