Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576420
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
GNE is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of malignant cells.
Specifications
GNE | |
Polyclonal | |
Unconjugated | |
GNE | |
2310066H07Rik; bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; DMRV; GLCNE; glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase; GNE; IBM2; ManAc kinase; N-acetylmannosamine kinase; N-acylmannosamine kinase; NM; Uae1; UDP-GlcNAc-2-epimerase; UDP-GlcNAc-2-epimerase/ManAc kinase; UDP-N-acetylglucosamine 2-epimerase (hydrolyzing); UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase; Uridine diphosphate-N-acetylglucosamine-2-epimerase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10020, 114711, 50798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O35826, Q91WG8, Q9Y223 | |
GNE | |
Recombinant fusion protein of human GNE. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction