Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576385
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction. Interacts with beta-1 and beta-2, but not with beta-3.
Specifications
GNG10 | |
Polyclonal | |
Unconjugated | |
GNG10 | |
AV006524; G protein subunit gamma 10; Gng10; GNGT10; guanine nucleotide binding protein (G protein), gamma 10; guanine nucleotide binding protein 10; guanine nucleotide binding protein gamma 10 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114119, 14700, 2790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P50151, Q9CXP8 | |
GNG10 | |
Recombinant fusion protein of human GNG10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction