Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gng12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529207
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (97%), Rat (95%), Xenopus laevis (81%), Pig (95%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. G proteins are composed of 3 units, alpha, beta and gamma.The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
| Gng12 | |
| Polyclonal | |
| Unconjugated | |
| GNG12 | |
| 2010305F15Rik; AA536815; AI115529; AI314170; AI842738; G protein subunit gamma 12; GNG12; G-protein gamma-12 subunit; guanine nucleotide binding protein (G protein), gamma 12; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14701, 55970 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9DAS9, Q9UBI6 | |
| GNG12 | |
| Full length human GNG12 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction