Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gng12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575620
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. G proteins are composed of 3 units, alpha, beta and gamma.The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
Gng12 | |
Polyclonal | |
Unconjugated | |
GNG12 | |
2010305F15Rik; AA536815; AI115529; AI314170; AI842738; G protein subunit gamma 12; GNG12; G-protein gamma-12 subunit; guanine nucleotide binding protein (G protein), gamma 12; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114120, 14701, 55970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9DAS9, Q9UBI6 | |
GNG12 | |
Synthetic peptide corresponding to residues in Human GNG12. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction