Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNG13 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300086MFV500
Description
GNG13 Polyclonal antibody specifically detects GNG13 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
GNG13 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
G gamma subunit, clone:h2-35, G(gamma)13, guanine nucleotide binding protein (G protein), gamma 13, guanine nucleotide binding protein 13, gamma, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-13, h2-35 | |
Produced in rabbits immunized with purified, recombinant Human GNG13 (Uniprot#: Q9P2W3; Met1-Cys64) | |
0.1 mL | |
Signal Transduction | |
51764 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction