Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561202
Description
Immunogen sequence: MEEWDVPQMK KEVESLKYQL AFQREMASKT IPELLKWIED GIPKDPFLNP DLMKNNPWVE KGKCTIL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
In mammals, G protein α, β and γ polypeptides are encoded by at least 16, 4 and 7 genes, respectively. Most interest in G proteins has been focused on their α subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Evidence, however, has established an important regulatory role for the βγ subunits. It is becoming increasingly clear that different G protein complexes expressed in different tissues carry structurally distinct members of the γ as well as the α and β subunits, and that preferential associations between members of subunit families increase G protein functional diversity.
Specifications
GNG13 | |
Polyclonal | |
Unconjugated | |
GNG13 | |
1500031D04Rik; AB030194; G gamma subunit, clone 2-35; G gamma subunit, clone:h2-35; G protein subunit gamma 13; G(gamma)13; Ggamma13; Gng13; guanine nucleotide binding protein (G protein), gamma 13; guanine nucleotide binding protein 13, gamma; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-13; h2-35 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51764 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2W3 | |
GNG13 | |
Recombinant protein corresponding to Human GNG13. Recombinant protein control fragment (Product #RP-98654). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction